(2RS)-2-Hydroxy-2-phenylacetic Acid (Mandelic Acid), Mikromol, LGC Standards
Catalog # 77692-901
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:90-64-2
- Description:(2RS)-2-Hydroxy-2-phenylacetic Acid (Mandelic Acid)
- Size:100 mg
- Formula:C₈H₈O₃
- Molecular Weight:152.15 g/mol
- Cat. No.:77692-901