$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Desethylhydroxychloroquine Dihydrochloride, Mikromol, LGC Standards
Desethylhydroxychloroquine Dihydrochloride, Mikromol, LGC Standards
77692-898
:
LGC Standards
New Product
:
Desethylhydroxychloroquine Dihydrochloride, Mikromol, LGC Standards
77692-898
:
LGC Standards
:
MM0764.02
New Product
:
CAS No.:
2732277-18-6
Description:
Desethylhydroxychloroquine Dihydrochloride
Size:
100 mg
Cat. No.:
77692-898