Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2,2-Dipropylpentanenitrile, Mikromol, LGC Standards
2,2-Dipropylpentanenitrile, Mikromol, LGC Standards
2,2-Dipropylpentanenitrile, Mikromol, LGC Standards
Catalog # 77692-897
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
2,2-Dipropylpentanenitrile, Mikromol, LGC Standards
Catalog # 77692-897
Supplier:
LGC Standards
Supplier Number:
MM0751.10
New Product
CAS Number:
Specifications
CAS No.:
5340-48-7
Description:
2,2-Dipropylpentanenitrile
Size:
100 mg
Cat. No.:
77692-897
Specifications
About this item
Product Details
Product Details