2-Bromo-1-(3-chlorophenyl)propan-1-one (2-Bromo-3'-chloropropiophenone), Mikromol, LGC Standards
77692-895
New Product
:
- CAS No.:34911-51-8
- Description:2-Bromo-1-(3-chlorophenyl)propan-1-one (2-Bromo-3'-chloropropiophenone)
- Size:100 mg
- Formula:C₉H₈BrClO
- Molecular Weight:247.516 g/mol
- Cat. No.:77692-895