$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
1-Phenyl-2-propanol, Mikromol, LGC Standards
1-Phenyl-2-propanol, Mikromol, LGC Standards
77692-891
:
LGC Standards
New Product
:
1-Phenyl-2-propanol, Mikromol, LGC Standards
77692-891
:
LGC Standards
:
MM0741.03
New Product
:
CAS No.:
698-87-3
Description:
1-Phenyl-2-propanol
Size:
100 mg
Formula:
C₆H₅CH₂CH(OH)CH₃
Molecular Weight:
136.19 g/mol
Cat. No.:
77692-891