(2S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic Acid (3,3',5'-Triiodo-L-Thyronine), Mikromol, LGC Standards
77692-887
New Product
:
- CAS No.:5817-39-0
- Description:(2S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic Acid (3,3',5'-Triiodo-L-Thyronine)
- Size:25 mg
- Formula:C₁₅H₁₂I₃NO₄
- Molecular Weight:650.97 g/mol
- Cat. No.:77692-887