Butylhydroxytoluene, Mikromol, LGC Standards
Catalog # 77692-885
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:128-37-0
- Description:Butylhydroxytoluene
- Size:250 mg
- Formula:C₁₅H₂₄O
- Molecular Weight:220.35 g/mol
- Cat. No.:77692-885