3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride), Mikromol, LGC Standards
77692-879
New Product
:
- CAS No.:31264-51-4
- Description:3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride)
- Size:100 mg
- Formula:C₁₁H₁₅ClO
- Molecular Weight:198.69 g/mol
- Cat. No.:77692-879