2,5,6-Triaminopyrimidin-4(1H)-one Sulphate, Mikromol, LGC Standards
Catalog # 77692-873
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:39267-74-8
- Description:2,5,6-Triaminopyrimidin-4(1H)-one Sulphate
- Size:100 mg
- Formula:C₄H₉N₅O₅S
- Molecular Weight:239.21 g/mol
- Cat. No.:77692-873