1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride), Mikromol, LGC Standards
Catalog # 77692-870
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:62-13-5
- Description:1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride)
- Size:100 mg
- Formula:C₉H₁₁NO₃·HCl·xH₂O
- Molecular Weight:217.65 g/mol (anhy)
- Cat. No.:77692-870