Adrenaline Tartrate, Mikromol, LGC Standards
Catalog # 77692-868
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:51-42-3
- Description:Adrenaline Tartrate
- Size:500 mg
- Formula:C₉H₁₃NO₃
- Molecular Weight:333.29 g/mol
- Cat. No.:77692-868