5H-Dibenzo[a,d][7]annulene (Dibenzocycloheptene), Mikromol, LGC Standards
Catalog # 77692-865
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:256-81-5
- Description:5H-Dibenzo[a,d][7]annulene (Dibenzocycloheptene)
- Size:50 mg
- Formula:C₁₅H₁₂
- Molecular Weight:192.26 g/mol
- Cat. No.:77692-865