3-[(2RS)-Piperidin-2-yl]pyridine ((RS)-Anabasine), Mikromol, LGC Standards
77692-834
New Product
:
- CAS No.:13078-04-1
- Description:3-[(2RS)-Piperidin-2-yl]pyridine ((RS)-Anabasine)
- Size:100 mg
- Formula:C₁₀H₁₄N₂
- Molecular Weight:162.236 g/mol
- Cat. No.:77692-834