4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride, Mikromol, LGC Standards
Catalog # 77692-805
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:28783-41-7
- Description:4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride
- Size:25 mg
- Formula:C₇H₁₀ClNS
- Molecular Weight:175.68 g/mol
- Cat. No.:77692-805