Order Entry
Puerto Rico
ContactUsLinkComponent
6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone), Mikromol, LGC Standards
6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone), Mikromol, LGC Standards
Catalog # 77692-798
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone), Mikromol, LGC Standards
Catalog # 77692-798
Supplier:  LGC Standards
Supplier Number:  MM0427.01
New Product
CAS Number:  

Specifications

  • CAS No.:
    2135-17-3
  • Description:
    6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone)
  • Size:
    100 mg
  • Formula:
    C₂₂H₂₈F₂O₅
  • Molecular Weight:
    410.46 g/mol
  • Cat. No.:
    77692-798

Specifications

About this item