2-[(4-Chlorophenyl)acetyl]benzoic Acid, Mikromol, LGC Standards
Catalog # 77692-767
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:53242-76-5
- Description:2-[(4-Chlorophenyl)acetyl]benzoic Acid
- Size:100 mg
- Formula:C₁₅H₁₁ClO₃
- Molecular Weight:274.7 g/mol
- Cat. No.:77692-767