(2R)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, Mikromol, LGC Standards
Catalog # 77692-716
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:22818-40-2
- Description:(2R)-2-Amino-2-(4-hydroxyphenyl)acetic Acid
- Size:100 mg
- Formula:C₈H₉NO₃
- Molecular Weight:167.16 g/mol
- Cat. No.:77692-716