Cyproterone Acetate, Mikromol, LGC Standards
Catalog # 77692-701
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:427-51-0
- Description:Cyproterone Acetate
- Size:250 mg
- Formula:C₂₄H₂₉ClO₄
- Molecular Weight:416.94 g/mol
- Cat. No.:77692-701