Order Entry
Puerto Rico
ContactUsLinkComponent
9-Fluoro-11beta,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Mikromol, LGC Standards
9-Fluoro-11beta,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Mikromol, LGC Standards
  77692-700
New Product
 :  
undefined
9-Fluoro-11beta,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Mikromol, LGC Standards
  77692-700
 :  MM0213.06
New Product
 :  

 

  • CAS No.:
    382-67-2
  • Description:
    9-Fluoro-11beta,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione
  • Size:
    100 mg
  • Formula:
    C₂₂H₂₉FO₄
  • Molecular Weight:
    376.47 g/mol
  • Cat. No.:
    77692-700