Beclometasone Dipropionate, Mikromol, LGC Standards
Catalog # 77692-688
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:5534-09-8
- Description:Beclometasone Dipropionate
- Size:250 mg
- Formula:C₂₈H₃₇ClO₇
- Molecular Weight:521.047 g/mol
- Cat. No.:77692-688