1-[Bis(4-fluorophenyl)methyl]piperazine, Mikromol, LGC Standards
Catalog # 77692-682
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:27469-60-9
- Description:1-[Bis(4-fluorophenyl)methyl]piperazine
- Size:100 mg
- Formula:C₁₇H₁₈F₂N₂
- Molecular Weight:288.34 g/mol
- Cat. No.:77692-682