Anisaldehyde, Mikromol, LGC Standards
Catalog # 77692-676
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:123-11-5
- Description:Anisaldehyde
- Size:100 mg
- Formula:C₈H₈O₂
- Molecular Weight:136.15 g/mol
- Cat. No.:77692-676