alpha-Demethylnaproxen, Mikromol, LGC Standards
Catalog # 77692-656
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:23981-47-7
- Description:alpha-Demethylnaproxen
- Size:100 mg
- Formula:C₁₃H₁₂O₃
- Molecular Weight:216.236 g/mol
- Cat. No.:77692-656