6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one, Mikromol, LGC Standards
Catalog # 77692-654
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:68559-60-4
- Description:6,7-Dihydrothieno[3,2-c]-pyridin-4(5H)-one
- Size:100 mg
- Formula:C₇H₇NOS
- Molecular Weight:153.20 g/mol
- Cat. No.:77692-654