1-Benzyl-1H-indazol-3-ol, Mikromol, LGC Standards
Catalog # 77692-645
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2215-63-6
- Description:1-Benzyl-1H-indazol-3-ol
- Size:100 mg
- Formula:C₁₄H₁₂N₂O
- Molecular Weight:224.26 g/mol
- Cat. No.:77692-645