4-Acetoxybenzoic Acid, Mikromol, LGC Standards
Catalog # 77692-643
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2345-34-8
- Description:4-Acetoxybenzoic Acid
- Size:100 mg
- Formula:C₉H₈O₄
- Molecular Weight:180.16 g/mol
- Cat. No.:77692-643