2-[(2-Hydroxybenzoyl)oxy]benzoic Acid (Salicylsalicylic Acid), Mikromol, LGC Standards
77692-641
New Product
:
- CAS No.:552-94-3
- Description:2-[(2-Hydroxybenzoyl)oxy]benzoic Acid (Salicylsalicylic Acid)
- Size:250 mg
- Formula:C₁₄H₁₀O₅
- Molecular Weight:258.23 g/mol
- Cat. No.:77692-641