Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2,6-Dimethylaniline, Mikromol, LGC Standards
2,6-Dimethylaniline, Mikromol, LGC Standards
77692-624
:
LGC Standards
New Product
:
2,6-Dimethylaniline, Mikromol, LGC Standards
77692-624
:
LGC Standards
:
MM0102.02
New Product
:
CAS No.:
87-62-7
Description:
2,6-Dimethylaniline
Size:
100 mg
Formula:
C₈H₁₁N
Molecular Weight:
121.18 g/mol
Cat. No.:
77692-624