Order Entry
Puerto Rico
ContactUsLinkComponent
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Mikromol, LGC Standards
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Mikromol, LGC Standards
Catalog # 77692-616
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole, Mikromol, LGC Standards
Catalog # 77692-616
Supplier:  LGC Standards
Supplier Number:  MM0095.04
New Product
CAS Number:  

Specifications

  • CAS No.:
    110374-16-8
  • Description:
    2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1H-benzimidazole
  • Size:
    100 mg
  • Formula:
    C₁₆H₁₇N₃O₂S
  • Molecular Weight:
    315.39 g/mol
  • Cat. No.:
    77692-616

Specifications

About this item