Order Entry
Puerto Rico
ContactUsLinkComponent
2-Chloro-6,7-dimethoxyquinazolin-4-amine, Mikromol, LGC Standards
2-Chloro-6,7-dimethoxyquinazolin-4-amine, Mikromol, LGC Standards
Supplier:  LGC Standards
New Product
Certificates
undefined
2-Chloro-6,7-dimethoxyquinazolin-4-amine, Mikromol, LGC Standards
Supplier:  LGC Standards
New Product

Specifications

  • CAS No.:
    23680-84-4
  • Formula:
    C₁₀H₁₀ClN₃O₂
  • Molecular Weight:
    239.66 g/mol

Specifications