1-(Furan-2-ylcarbonyl)piperazine, Mikromol, LGC Standards
Catalog # 77692-599
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:40172-95-0
- Description:1-(Furan-2-ylcarbonyl)piperazine
- Size:100 mg
- Formula:C₉H₁₂N₂O₂
- Molecular Weight:180.21 g/mol
- Cat. No.:77692-599