6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine, Mikromol, LGC Standards
77692-598
New Product
:
- CAS No.:60547-97-9
- Description:6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine
- Size:100 mg
- Formula:C₁₄H₁₉N₅O₂
- Molecular Weight:289.333 g/mol
- Cat. No.:77692-598