5H-Dibenzo[b,f]azepine (Iminostilbene), Mikromol, LGC Standards
Catalog # 77692-578
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:256-96-2
- Description:5H-Dibenzo[b,f]azepine (Iminostilbene)
- Size:100 mg
- Formula:C₁₄H₁₁N
- Molecular Weight:193.25 g/mol
- Cat. No.:77692-578