10,11-Dihydro-5H-dibenzo[b,f]azepine (Iminodibenzyl), Mikromol, LGC Standards
Catalog # 77692-577
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:494-19-9
- Description:10,11-Dihydro-5H-dibenzo[b,f]azepine (Iminodibenzyl)
- Size:100 mg
- Formula:C₁₄H₁₃N
- Molecular Weight:195.27 g/mol
- Cat. No.:77692-577