Bromhexine Hydrochloride, Mikromol, LGC Standards
Catalog # 77692-564
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:611-75-6
- Description:Bromhexine Hydrochloride
- Size:250 mg
- Formula:C₁₄H₂₀Br₂N₂
- Molecular Weight:412.59 g/mol
- Cat. No.:77692-564