10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane), Mikromol, LGC Standards
77692-558
New Product
:
- CAS No.:833-48-7
- Description:10,11-Dihydro-5H-dibenzo[a,d]cycloheptene (Dibenzosuberane)
- Size:100 mg
- Formula:C₁₅H₁₄
- Molecular Weight:194.28 g/mol
- Cat. No.:77692-558