Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Amitriptyline N-Oxide Hydrochloride, Mikromol, LGC Standards
Amitriptyline N-Oxide Hydrochloride, Mikromol, LGC Standards
Amitriptyline N-Oxide Hydrochloride, Mikromol, LGC Standards
Catalog # 77692-557
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Amitriptyline N-Oxide Hydrochloride, Mikromol, LGC Standards
Catalog # 77692-557
Supplier:
LGC Standards
Supplier Number:
MM0062.12
New Product
CAS Number:
Specifications
CAS No.:
4290-60-2
Description:
Amitriptyline N-Oxide Hydrochloride
Size:
10 mg
Cat. No.:
77692-557
Specifications
About this item
Product Details
Product Details