5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-ol, Mikromol, LGC Standards
77692-556
New Product
:
- CAS No.:1159-03-1
- Description:5-[3-(Dimethylamino)propyl]-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-ol
- Size:100 mg
- Formula:C₂₀H₂₅NO
- Molecular Weight:295.42 g/mol
- Cat. No.:77692-556