Order Entry
Puerto Rico
ContactUsLinkComponent
4-Amino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (Ampyrone), Mikromol, LGC Standards
4-Amino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (Ampyrone), Mikromol, LGC Standards
Catalog # 77692-547
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
4-Amino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (Ampyrone), Mikromol, LGC Standards
Catalog # 77692-547
Supplier:  LGC Standards
Supplier Number:  MM0052.04
New Product
CAS Number:  

Specifications

  • CAS No.:
    83-07-8
  • Description:
    4-Amino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (Ampyrone)
  • Size:
    100 mg
  • Formula:
    C₁₁H₁₃N₃O
  • Molecular Weight:
    203.24 g/mol
  • Cat. No.:
    77692-547

Specifications

About this item