Order Entry
Puerto Rico
ContactUsLinkComponent
4-Formylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (4-Formylaminophenazone), Mikromol, LGC Standards
4-Formylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (4-Formylaminophenazone), Mikromol, LGC Standards
Catalog # 77692-546
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
4-Formylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (4-Formylaminophenazone), Mikromol, LGC Standards
Catalog # 77692-546
Supplier:  LGC Standards
Supplier Number:  MM0052.02
New Product
CAS Number:  

Specifications

  • CAS No.:
    1672-58-8
  • Description:
    4-Formylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one (4-Formylaminophenazone)
  • Size:
    100 mg
  • Formula:
    C₁₂H₁₃N₃O₂
  • Molecular Weight:
    231.26 g/mol
  • Cat. No.:
    77692-546

Specifications

About this item