5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one (Edaravone), Mikromol, LGC Standards
77692-530
New Product
:
- CAS No.:89-25-8
- Description:5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one (Edaravone)
- Size:100 mg
- Formula:C₁₀H₁₀N₂O
- Molecular Weight:174.2 g/mol
- Cat. No.:77692-530