4-(4-Chlorophenyl)piperidin-4-ol, Mikromol, LGC Standards
Catalog # 77692-504
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:39512-49-7
- Description:4-(4-Chlorophenyl)piperidin-4-ol
- Size:25 mg
- Formula:C₁₁H₁₄ClNO
- Molecular Weight:211.69 g/mol
- Cat. No.:77692-504