Order Entry
Puerto Rico
ContactUsLinkComponent
2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine (Cimetidine Sulphoxide), Mikromol, LGC Standards
2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine (Cimetidine Sulphoxide), Mikromol, LGC Standards
Catalog # 77692-499
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine (Cimetidine Sulphoxide), Mikromol, LGC Standards
Catalog # 77692-499
Supplier:  LGC Standards
Supplier Number:  MM0020.04-0025
New Product
CAS Number:  

Specifications

  • CAS No.:
    54237-72-8
  • Description:
    2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine (Cimetidine Sulphoxide)
  • Size:
    25 mg
  • Formula:
    C₁₀H₁₆N₆OS
  • Molecular Weight:
    268.3386 g/mol
  • Cat. No.:
    77692-499

Specifications

About this item