Order Entry
Puerto Rico
ContactUsLinkComponent
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat), Mikromol, LGC Standards
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat), Mikromol, LGC Standards
Catalog # 77692-476
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat), Mikromol, LGC Standards
Catalog # 77692-476
Supplier:  LGC Standards
Supplier Number:  MM0010.01
New Product
CAS Number:  

Specifications

  • CAS No.:
    76420-72-9
  • Description:
    (2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat)
  • Size:
    100 mg
  • Formula:
    C₁₈H₂₈N₂O₇
  • Molecular Weight:
    384.42 g/mol
  • Cat. No.:
    77692-476

Specifications

About this item