Diclofenac Diethylamine (Diethylammonium Diclofenac), Mikromol, LGC Standards
Catalog # 77692-472
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:78213-16-8
- Description:Diclofenac Diethylamine (Diethylammonium Diclofenac)
- Size:250 mg
- Formula:C₁₈H₂₂Cl₂N₂O₂
- Molecular Weight:369.28 g/mol
- Cat. No.:77692-472