Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
Cinacalcet Hydrochloride, LoGiCal, LGC Standards
Cinacalcet Hydrochloride, LoGiCal, LGC Standards
77692-438
:
LGC Standards
New Product
:
Cinacalcet Hydrochloride, LoGiCal, LGC Standards
77692-438
:
LGC Standards
:
LGCFOR3370.00
New Product
:
CAS No.:
364782-34-3
Description:
Cinacalcet Hydrochloride
Size:
10 mg
Formula:
C₂₂H₂₂F₃N·HCl
Molecular Weight:
393.88 g/mol
Cat. No.:
77692-438