$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Diazoxide, LoGiCal, LGC Standards
Diazoxide, LoGiCal, LGC Standards
:
LGC Standards
New Product
Diazoxide, LoGiCal, LGC Standards
:
LGC Standards
New Product
CAS No.:
364-98-7
Formula:
C₈H₇ClN₂O₂S
Molecular Weight:
230.666 g/mol