Clonidine Hydrochloride, LoGiCal, LGC Standards
Catalog # 77692-411
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:4205-91-8
- Description:Clonidine Hydrochloride
- Size:10 mg
- Formula:C₉H₉Cl₂N₃
- Molecular Weight:266.55 g/mol
- Cat. No.:77692-411