Chlortetracycline Hydrochloride, LoGiCal, LGC Standards
Catalog # 77692-402
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:64-72-2
- Description:Chlortetracycline Hydrochloride
- Size:10 mg
- Formula:C₂₂H₂₃ClN₂O₈·HCl
- Molecular Weight:515.34 g/mol
- Cat. No.:77692-402