9-Hydroxyrisperidone, LoGiCal, LGC Standards
Catalog # 77692-397
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:144598-75-4
- Description:9-Hydroxyrisperidone
- Size:10 mg
- Formula:C₂₃H₂₇FN₄O₃
- Molecular Weight:426.49 g/mol
- Cat. No.:77692-397